Please login to your online account to display your discounted pricing

5-methyl-1-[3-(trifluoromethyl)phenyl]-1H-1,2,3-triazole-4-carboxylic acid, Maybridge


Quantity 1g

Manufacturer: fisher scientific company  CD04550DA

 View more versions of this product

Catalog No. CD04550DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 271.2
Quantity 1g
Assay 97%
Assay Percent Range 97%
Chemical Name or Material 5-Methyl-1-[3-(trifluoromethyl)phenyl]-1H-1, 2, 3-triazole-4-carboxylic acid
Health Hazard 1 GHS Signal Word: Warning
Molecular Formula C11H8F3N3O2
CAS 499771-21-0

Specifications

Chemical Name or Material 5-Methyl-1-[3-(trifluoromethyl)phenyl]-1H-1, 2, 3-triazole-4-carboxylic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H8F3N3O2
Formula Weight 271.2
Accessories

GHS Signal Word: Warning

SDS & Safety Documents