Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Methyl-1,3,4-oxadiazole-2-methylamine oxalate, 99%

Chemical Identifiers

Chemical Name or Material 5-Methyl-1,3,4-oxadiazole-2-methylamine oxalate
Molecular Formula C4H7N3O·C2H2O4
Formula Weight 203.15
MDL Number MFCD07781053
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5016203

  SDS
alfa aesar™
H5016203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH50162MD

  SDS
alfa aesar™
H50162MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 203.15
Molecular Formula C4H7N3O·C2H2O4
Health Hazard 1 Danger
Quantity 1g
Chemical Name or Material 5-Methyl-1,3,4-oxadiazole-2-methylamine oxalate
Health Hazard 3 GHS P Statement
P280-P301+P310-P361-P312-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD07781053
Health Hazard 2 GHS H Statement
H301-H311
Toxic if swallowed.
Toxic in contact with skin.
Assay 99%

Applications
5-Methyl-1,3,4-oxadiazole-2-methylamine oxalate is used as pharmaceutical intermediate.

Notes
Air sensitive. Store away from oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P280-P301+P310-P361-P312-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311
Toxic if swallowed.
Toxic in contact with skin.