missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Methyl-1,10-phenanthroline Hydrate [for Colorimetric Determination of Iron], TCI America™

$55.95 - $258.41

Chemical Identifiers

CAS 3002-78-6
Molecular Formula C13H10N2
Molecular Weight (g/mol) 194.237
MDL Number MFCD00004982
InChI Key UJAQYOZROIFQHO-UHFFFAOYSA-N
Synonym 1,10-phenanthroline, 5-methyl, 1,10-phenanthroline,5-methyl, 5-methyl-1,10 phenanthroline, 5-methyl-1,10-phenanthroline, 5-methylphenanthroline, 5-methylpyridino 3,2-h quinoline, acmc-1agcx, chembl98682, monohydrate, ujaqyozroifqho-uhfffaoysa
PubChem CID 72791
IUPAC Name 5-methyl-1,10-phenanthroline
SMILES CC1=C2C=CC=NC2=C3C(=C1)C=CC=N3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M03001G View Documents Tci America
M03001G
1g Each for $258.41
 
M0300100MG View Documents Tci America
M0300100MG
100mg Each for $55.95
 
Specifications

Chemical Identifiers

3002-78-6
194.237
UJAQYOZROIFQHO-UHFFFAOYSA-N
72791
CC1=C2C=CC=NC2=C3C(=C1)C=CC=N3
C13H10N2
MFCD00004982
1,10-phenanthroline, 5-methyl, 1,10-phenanthroline,5-methyl, 5-methyl-1,10 phenanthroline, 5-methyl-1,10-phenanthroline, 5-methylphenanthroline, 5-methylpyridino 3,2-h quinoline, acmc-1agcx, chembl98682, monohydrate, ujaqyozroifqho-uhfffaoysa
5-methyl-1,10-phenanthroline

Specifications

Undesignated
1g
C13H10N2
1,10-phenanthroline, 5-methyl, 1,10-phenanthroline,5-methyl, 5-methyl-1,10 phenanthroline, 5-methyl-1,10-phenanthroline, 5-methylphenanthroline, 5-methylpyridino 3,2-h quinoline, acmc-1agcx, chembl98682, monohydrate, ujaqyozroifqho-uhfffaoysa
CC1=C2C=CC=NC2=C3C(=C1)C=CC=N3
194.237
194.24
5-Methyl-1,10-phenanthroline Hydrate [for Colorimetric Determination of Iron]
113°C
3002-78-6
MFCD00004982
UJAQYOZROIFQHO-UHFFFAOYSA-N
5-methyl-1,10-phenanthroline
72791
Crystal-Powder at 20°C
Safety and Handling

Safety and Handling

TSCA : Yes