missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Methyl-1,10-phenanthroline Hydrate [for Colorimetric Determination of Iron], TCI America™
Supplier: TCI America M03001G
Specifications
5-Methyl-1,10-phenanthroline Hydrate [for Colorimetric Determination of Iron] | |
113°C | |
MFCD00004982 | |
1,10-phenanthroline, 5-methyl, 5-methylphenanthroline, chembl98682, 1,10-phenanthroline,5-methyl, acmc-1agcx, 5-methyl-1,10 phenanthroline, ujaqyozroifqho-uhfffaoysa, 5-methylpyridino 3,2-h quinoline, 5-methyl-1,10-phenanthroline, monohydrate | |
CC1=C2C=CC=NC2=C3C(=C1)C=CC=N3 | |
194.237 | |
194.24 |
3002-78-6 | |
C13H10N2 | |
1 g | |
UJAQYOZROIFQHO-UHFFFAOYSA-N | |
5-methyl-1,10-phenanthroline | |
72791 | |
Crystalline Powder |
Chemical Identifiers
3002-78-6 | |
194.237 | |
UJAQYOZROIFQHO-UHFFFAOYSA-N | |
72791 | |
CC1=C2C=CC=NC2=C3C(=C1)C=CC=N3 |
C13H10N2 | |
MFCD00004982 | |
1,10-phenanthroline, 5-methyl, 5-methylphenanthroline, chembl98682, 1,10-phenanthroline,5-methyl, acmc-1agcx, 5-methyl-1,10 phenanthroline, ujaqyozroifqho-uhfffaoysa, 5-methylpyridino 3,2-h quinoline, 5-methyl-1,10-phenanthroline, monohydrate | |
5-methyl-1,10-phenanthroline |
Safety and Handling
TSCA : Yes