Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Methoxyresorcinol, 95%

Chemical Identifiers

Chemical Name or Material 5-Methoxyresorcinol
CAS 2174-64-3
Formula Weight 140.14
MDL Number MFCD00002285
Beilstein 1423578
Synonym 3,5-Dihydroxyanisole; 3,5-Dihydroxymethoxybenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6199603

  SDS
alfa aesar™
H6199603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6199606

  SDS
alfa aesar™
H6199606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH61996MD

  SDS
alfa aesar™
H61996MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 140.14
Boiling Point 188° to 189°C (12mmHg)
Melting Point 78° to 80°C
Chemical Name or Material 5-Methoxyresorcinol
Physical Form Solid
Assay 95%
MDL Number MFCD00002285
CAS 2174-64-3
Color Yellow
Beilstein 1423578
Synonym 3,5-Dihydroxyanisole; 3,5-Dihydroxymethoxybenzene
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
5-Methoxyresorcinol was used in synthesis of isorobustin, substituted linear and angular benzofurocoumarins.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.