Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Methoxymethyl-2-furoic acid, 99%

$94.93 - $337.51

Chemical Identifiers

Chemical Name or Material 5-Methoxymethyl-2-furoic acid
CAS 1917-60-8
Molecular Formula C7H8O4
Formula Weight 156.14
MDL Number MFCD04367609
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5031403

  SDS
alfa aesar™
H5031403
1g Each for $337.51

AAH50314MD

  SDS
alfa aesar™
H50314MD
250mg Each for $94.93
Description & Specifications

Specifications

Formula Weight 156.14
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 1917-60-8
Quantity 1g
Chemical Name or Material 5-Methoxymethyl-2-furoic acid
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 99%
Health Hazard 1 Warning
Molecular Formula C7H8O4
MDL Number MFCD04367609
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.