missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Methoxyisatin, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 39755-95-8 |
---|---|
Molecular Formula | C9H7NO3 |
Molecular Weight (g/mol) | 177.16 |
MDL Number | MFCD00169023 |
InChI Key | DMHGXMPXHPOXBF-UHFFFAOYSA-N |
Synonym | 5-methoxyisatin, 5-methoxyindoline-2,3-dione, 5-methoxy-2,3-indolinedione, 1h-indole-2,3-dione, 5-methoxy, 5-methoxy-indole-2,3-dione, 5-methoxy isatin, 5-methoxyindole-2,3-dione, 5-methoxy-2,3-dihydro-1h-indole-2,3-dione, indole-2,3-dione, 5-methoxy, 1h-indole-2,3-dione, 5-methoxy-9ci |
PubChem CID | 38333 |
IUPAC Name | 5-methoxy-1H-indole-2,3-dione |
SMILES | COC1=CC=C2NC(=O)C(=O)C2=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC448830050
|
Thermo Scientific Chemicals
448830050 |
5g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC448830250
|
Thermo Scientific Chemicals
448830250 |
25g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC448831000
|
Thermo Scientific Chemicals
448831000 |
100g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
39755-95-8 | |
177.16 | |
DMHGXMPXHPOXBF-UHFFFAOYSA-N | |
38333 | |
COC1=CC=C2NC(=O)C(=O)C2=C1 |
C9H7NO3 | |
MFCD00169023 | |
5-methoxyisatin, 5-methoxyindoline-2,3-dione, 5-methoxy-2,3-indolinedione, 1h-indole-2,3-dione, 5-methoxy, 5-methoxy-indole-2,3-dione, 5-methoxy isatin, 5-methoxyindole-2,3-dione, 5-methoxy-2,3-dihydro-1h-indole-2,3-dione, indole-2,3-dione, 5-methoxy, 1h-indole-2,3-dione, 5-methoxy-9ci | |
5-methoxy-1H-indole-2,3-dione |
Specifications
39755-95-8 | |
Glass bottle | |
MFCD00169023 | |
5-methoxyisatin, 5-methoxyindoline-2,3-dione, 5-methoxy-2,3-indolinedione, 1h-indole-2,3-dione, 5-methoxy, 5-methoxy-indole-2,3-dione, 5-methoxy isatin, 5-methoxyindole-2,3-dione, 5-methoxy-2,3-dihydro-1h-indole-2,3-dione, indole-2,3-dione, 5-methoxy, 1h-indole-2,3-dione, 5-methoxy-9ci | |
COC1=CC=C2NC(=O)C(=O)C2=C1 | |
177.16 | |
177.16 | |
5-Methoxyisatin |
96% min. (GC) | |
C9H7NO3 | |
5g | |
DMHGXMPXHPOXBF-UHFFFAOYSA-N | |
5-methoxy-1H-indole-2,3-dione | |
38333 | |
97% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
RUO â Research Use Only