missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Methoxyindole-3-carboxylic acid, 97%

$39.78 - $467.26

Chemical Identifiers

CAS 10242-01-0
Molecular Formula C10H9NO3
Molecular Weight (g/mol) 191.186
MDL Number MFCD03265451
InChI Key RVVSEZGJCOAUED-UHFFFAOYSA-N
Synonym 5-methoxyindole-3-carboxylic acid, 5-methoxy-3-indolecarboxylic acid, 5-methoxy-3-carboxyindole, 1h-indole-3-carboxylic acid, 5-methoxy, pubchem7984, zlchem 1276, 1h-indole-3-carboxylicacid, 5-methoxy, 5-methoxy-3-indolecarboxylic acid, 5-methoxyindole-3-carboxylic acid;, 5-methoxyindole-3-carboxylic acid 5-methoxy-1h-indazole-3-carboxylic acid
PubChem CID 259191
IUPAC Name 5-methoxy-1H-indole-3-carboxylic acid
SMILES COC1=CC2=C(C=C1)NC=C2C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64722MD View Documents Alfa Aesar™
H64722MD
250mg Each for $39.78
 
AAH6472203 View Documents Alfa Aesar™
H6472203
1g Each for $117.66
 
AAH6472206 View Documents Alfa Aesar™
H6472206
5g Each for $467.26
 
Specifications

Chemical Identifiers

10242-01-0
191.186
RVVSEZGJCOAUED-UHFFFAOYSA-N
259191
COC1=CC2=C(C=C1)NC=C2C(=O)O
C10H9NO3
MFCD03265451
5-methoxyindole-3-carboxylic acid, 5-methoxy-3-indolecarboxylic acid, 5-methoxy-3-carboxyindole, 1h-indole-3-carboxylic acid, 5-methoxy, pubchem7984, zlchem 1276, 1h-indole-3-carboxylicacid, 5-methoxy, 5-methoxy-3-indolecarboxylic acid, 5-methoxyindole-3-carboxylic acid;, 5-methoxyindole-3-carboxylic acid 5-methoxy-1h-indazole-3-carboxylic acid
5-methoxy-1H-indole-3-carboxylic acid

Specifications

Red
10242-01-0
MFCD03265451
5-methoxyindole-3-carboxylic acid, 5-methoxy-3-indolecarboxylic acid, 5-methoxy-3-carboxyindole, 1h-indole-3-carboxylic acid, 5-methoxy, pubchem7984, zlchem 1276, 1h-indole-3-carboxylicacid, 5-methoxy, 5-methoxy-3-indolecarboxylic acid, 5-methoxyindole-3-carboxylic acid;, 5-methoxyindole-3-carboxylic acid 5-methoxy-1h-indazole-3-carboxylic acid
COC1=CC2=C(C=C1)NC=C2C(=O)O
191.186
191.19
97%
250mg
C10H9NO3
475711
RVVSEZGJCOAUED-UHFFFAOYSA-N
5-methoxy-1H-indole-3-carboxylic acid
259191
Crystalline
5-Methoxyindole-3-carboxylic acid
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C