Please login to your online account to display your discounted pricing

5-Methoxyindole-3-carboxaldehyde 99+%, ACROS Organics™

$129.68 - $129.68

Chemical Identifiers

Chemical Name or Material 5-Methoxyindole-3-carboxaldehyde
Name Note 99+%
CAS 10601-19-1
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C10H9NO2
Formula Weight 175.19
MDL Number MFCD00005623
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC125890010

  SDS
thermo fisher scientific
125890010
Glass bottle 1g Each for $129.68

AC125890050

  SDS
thermo fisher scientific
125890050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC125891000

  SDS
thermo fisher scientific
AC125891000
Glass bottle 100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder or Needles
Assay ≥99.0%
Formula Weight 175.19
Color Yellow to beige
Quantity 1g
Beilstein 21, II, 424
EINECS Number 234-220-5
Melting Point 180° to 184°C
CAS Min % 99.0
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Chemical Name or Material 5-Methoxyindole-3-carboxaldehyde
Name Note 99+%
Assay Percent Range 99+%
Molecular Formula C10H9NO2
MDL Number MFCD00005623
CAS 10601-19-1
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.