missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Methoxyindole-2-carboxylic acid, 99%, Thermo Scientific™
Chemical Identifiers
CAS | 4382-54-1 |
---|---|
Molecular Formula | C10H9NO3 |
Molecular Weight (g/mol) | 191.19 |
MDL Number | MFCD00005614 |
InChI Key | YEBJVSLNUMZXRJ-UHFFFAOYSA-N |
Synonym | 5-methoxyindole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 5-methoxy, 5-methoxy-2-indolecarboxylic acid, indole-2-carboxylic acid, 5-methoxy, acide methoxy-5 indole carboxylique-2, acide methoxy-5 indole carboxylique-2 french, 5-methoxyindol-2-carboxylic acid, 5-methoxy-1h-indole-2-carboxylicacid, pubchem1702, acmc-209jvj |
PubChem CID | 20401 |
IUPAC Name | 5-methoxy-1H-indole-2-carboxylic acid |
SMILES | COC1=CC=C2NC(=CC2=C1)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC125900100
|
Thermo Scientific Chemicals
125900100 |
10g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
4382-54-1 | |
191.19 | |
YEBJVSLNUMZXRJ-UHFFFAOYSA-N | |
20401 | |
COC1=CC=C2NC(=CC2=C1)C(O)=O |
C10H9NO3 | |
MFCD00005614 | |
5-methoxyindole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 5-methoxy, 5-methoxy-2-indolecarboxylic acid, indole-2-carboxylic acid, 5-methoxy, acide methoxy-5 indole carboxylique-2, acide methoxy-5 indole carboxylique-2 french, 5-methoxyindol-2-carboxylic acid, 5-methoxy-1h-indole-2-carboxylicacid, pubchem1702, acmc-209jvj | |
5-methoxy-1H-indole-2-carboxylic acid |
Specifications
4382-54-1 | |
100.0 | |
99% | |
C10H9NO3 | |
10g | |
5-methoxyindole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 5-methoxy, 5-methoxy-2-indolecarboxylic acid, indole-2-carboxylic acid, 5-methoxy, acide methoxy-5 indole carboxylique-2, acide methoxy-5 indole carboxylique-2 french, 5-methoxyindol-2-carboxylic acid, 5-methoxy-1h-indole-2-carboxylicacid, pubchem1702, acmc-209jvj | |
YEBJVSLNUMZXRJ-UHFFFAOYSA-N | |
5-methoxy-1H-indole-2-carboxylic acid | |
20401 | |
99% |
98.5 | |
Authentic | |
Glass bottle | |
MFCD00005614 | |
22, II, 169 | |
Solubility in water: slightly soluble in water. Other solubilities: soluble in acetone | |
COC1=CC=C2NC(=CC2=C1)C(O)=O | |
191.19 | |
191.19 | |
5-Methoxyindole-2-carboxylic acid, 99% |
Safety and Handling
EINECSNumber : 224-487-6
RTECSNumber : NL6005000
RUO â Research Use Only