missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Methoxyflavone 98.0+%, TCI America™

$238.66 - $768.63

Chemical Identifiers

CAS 42079-78-7
Molecular Formula C16H12O3
Molecular Weight (g/mol) 252.269
MDL Number MFCD00016942
InChI Key XRQSPUXANRGDAV-UHFFFAOYSA-N
Synonym 4h-1-benzopyran-4-one, 5-methoxy-2-phenyl, 5-methoxy-2-phenyl-4-benzopyrone, 5-methoxy-2-phenyl-4h-chromen-4-one, 5-methoxy-2-phenyl-chromen-4-one, 5-methoxyflavone, acmc-20am2h, bidd:er0473, chembl16685, d0v5hf
PubChem CID 94525
IUPAC Name 5-methoxy-2-phenylchromen-4-one
SMILES COC1=CC=CC2=C1C(=O)C=C(O2)C3=CC=CC=C3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M23651G View Documents Tci America
M23651G
1g Each for $238.66
 
M23655G View Documents Tci America
M23655G
5g Each for $768.63
 
Specifications

Chemical Identifiers

42079-78-7
252.269
XRQSPUXANRGDAV-UHFFFAOYSA-N
94525
COC1=CC=CC2=C1C(=O)C=C(O2)C3=CC=CC=C3
C16H12O3
MFCD00016942
4h-1-benzopyran-4-one, 5-methoxy-2-phenyl, 5-methoxy-2-phenyl-4-benzopyrone, 5-methoxy-2-phenyl-4h-chromen-4-one, 5-methoxy-2-phenyl-chromen-4-one, 5-methoxyflavone, acmc-20am2h, bidd:er0473, chembl16685, d0v5hf
5-methoxy-2-phenylchromen-4-one

Specifications

White-Yellow
1g
C16H12O3
4h-1-benzopyran-4-one, 5-methoxy-2-phenyl, 5-methoxy-2-phenyl-4-benzopyrone, 5-methoxy-2-phenyl-4h-chromen-4-one, 5-methoxy-2-phenyl-chromen-4-one, 5-methoxyflavone, acmc-20am2h, bidd:er0473, chembl16685, d0v5hf
COC1=CC=CC2=C1C(=O)C=C(O2)C3=CC=CC=C3
252.269
252.27
≥98.0% (GC)
132°C
42079-78-7
MFCD00016942
XRQSPUXANRGDAV-UHFFFAOYSA-N
5-methoxy-2-phenylchromen-4-one
94525
Crystal-Powder at 20°C
5-Methoxyflavone
Safety and Handling

Safety and Handling

TSCA : No