Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Methoxyflavone, 97%

$115.14 - $484.88

Chemical Identifiers

Chemical Name or Material 5-Methoxyflavone
CAS 42079-78-7
Molecular Formula C16H12O3
Formula Weight 252.27
MDL Number MFCD00016942
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2370002

 
alfa aesar™
B2370002
0.5g Each for $115.14

AAB2370088

 
alfa aesar™
B2370088
2.5g Each for $484.88
Description & Specifications

Specifications

Formula Weight 252.27
Chemical Name or Material 5-Methoxyflavone
Assay 97%
Molecular Formula C16H12O3
MDL Number MFCD00016942
CAS 42079-78-7
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 128° to 131°C
Quantity 0.5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.