Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Methoxybenzoxazole-2-thiol

Chemical Identifiers

Chemical Name or Material 5-Methoxybenzoxazole-2-thiol
CAS 49559-83-3
Molecular Formula C8H7NO2S
Formula Weight 181.21
MDL Number MFCD03030041
Synonym 2-Mercapto-5-methoxybenzo[d]oxazole; 5-Methoxy-2(3H)-benzoxazolethione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ6473009

 
alfa aesar™
J6473009
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 181.21
Chemical Name or Material 5-Methoxybenzoxazole-2-thiol
MDL Number MFCD03030041
Physical Form Powder
Molecular Formula C8H7NO2S
CAS 49559-83-3
Synonym 2-Mercapto-5-methoxybenzo[d]oxazole; 5-Methoxy-2(3H)-benzoxazolethione
Sensitivity Moisture and Light Sensitive
Health Hazard 2 GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
Color White to Off-white
Melting Point 218° to 223°C
Quantity 10g
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

2° to 8°C