Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Methoxybenzimidazole, 99%

$34.35 - $510.80

Chemical Identifiers

Chemical Name or Material 5-Methoxybenzimidazole
CAS 4887-80-3
Molecular Formula C8H8N2O
Formula Weight 148.17
MDL Number MFCD00272526
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2431303

  SDS
alfa aesar™
B2431303
1g Each for $34.35

AAB2431306

  SDS
alfa aesar™
B2431306
5g Each for $129.14

AAB2431314

  SDS
alfa aesar™
B2431314
25g Each for $510.80
Description & Specifications

Specifications

Formula Weight 148.17
Chemical Name or Material 5-Methoxybenzimidazole
Assay 99%
Molecular Formula C8H8N2O
MDL Number MFCD00272526
CAS 4887-80-3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 121° to 123°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.