Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Methoxy-2-(trifluoromethyl)pyridine, 97%

$81.27 - $270.92

Chemical Identifiers

Chemical Name or Material 5-Methoxy-2-(trifluoromethyl)pyridine
CAS 216766-13-1
Molecular Formula C7H6F3NO
Formula Weight 177.13
MDL Number MFCD09864699
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3258603

 
alfa aesar™
H3258603
1g Each for $81.27

AAH3258606

 
alfa aesar™
H3258606
5g Each for $270.92
Description & Specifications

Specifications

Formula Weight 177.13
Chemical Name or Material 5-Methoxy-2-(trifluoromethyl)pyridine
Molecular Formula C7H6F3NO
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Assay 97%
MDL Number MFCD09864699
CAS 216766-13-1
Quantity 1g
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.