Learn More
5-Methoxy-2-nitrophenol, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 335440010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
5-Methoxy-2-nitrophenol | |
97.5 | |
90°C to 94°C | |
Authentic | |
Glass bottle | |
MFCD00100932 | |
2-hydroxy-4-methoxynitrobenzene, phenol, 5-methoxy-2-nitro, 2-nitro-5-methoxyphenol, 3-hydroxy-4-nitroanisole, 5-methoxy-2-nitro-phenol, phenol,5-methoxy-2-nitro, pubchem22230, 3-methoxy-6-nitrophenol, 2-hydroxy4-methoxynitrobenzene, 5-methoxy-2-nitrophenol | |
NRTULWPODYLFOJ-UHFFFAOYSA-N | |
5-methoxy-2-nitrophenol | |
219635 | |
98% |
704-14-3 | |
100.0 | |
Yellow | |
97.5% min. (GC) | |
C7H7NO4 | |
1 g | |
Solubility in water: .. Other solubilities: soluble in acetone,chloroform,ethyl alcohol | |
COC1=CC=C(C(O)=C1)[N+]([O-])=O | |
169.14 | |
169.14 | |
Crystals or Powder |
Chemical Identifiers
704-14-3 | |
169.14 | |
NRTULWPODYLFOJ-UHFFFAOYSA-N | |
5-methoxy-2-nitrophenol |
C7H7NO4 | |
MFCD00100932 | |
219635 | |
COC1=CC=C(C(O)=C1)[N+]([O-])=O |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
GHS Signal Word: Danger
RUO – Research Use Only