Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Methoxy-2-nitrobenzoic acid, 97%

$61.57 - $61.57

Chemical Identifiers

Chemical Name or Material 5-Methoxy-2-nitrobenzoic acid
CAS 1882-69-5
Molecular Formula C8H7NO5
Formula Weight 197.15
MDL Number MFCD00151836
Beilstein 1877657
Synonym 6-Nitro-m-anisic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1334606

 
alfa aesar™
L1334606
5g Each for $61.57
Description & Specifications

Specifications

Formula Weight 197.15
Chemical Name or Material 5-Methoxy-2-nitrobenzoic acid
MDL Number MFCD00151836
CAS 1882-69-5
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
Molecular Formula C8H7NO5
Beilstein 1877657
Melting Point 130° to 135°C
Synonym 6-Nitro-m-anisic acid
Quantity 5g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.