missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Methoxy-2-methylindole, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC125940010

 View more versions of this product

Catalog No. AC125940010



Description

Description

Specifications

Specifications

5-Methoxy-2-methylindole
Glass bottle
85°C to 88°C
99+%
99.0
99% min. (GC)
MFCD00005620
1h-indole, 5-methoxy-2-methyl, 2-methyl-5-methoxy indole, 2-methyl-5-methoxyindole, 2-methyl-5-methoxylindole, 5-methoxy-2 methylindole, 5-methoxy-2-methylindole, acmc-1buiu, indole, 5-methoxy-2-methyl, maybridge1_002343, pubchem7236
CC1=CC2=C(N1)C=CC(=C2)OC
161.2
161.2
99+%
Authentic
Beige to White
1g
1076-74-0
100.0
C10H11NO
21, II, 47
VSWGLJOQFUMFOQ-UHFFFAOYSA-N
5-methoxy-2-methyl-1H-indole
70642
Crystalline Powder or Crystals

Chemical Identifiers

1076-74-0
161.2
VSWGLJOQFUMFOQ-UHFFFAOYSA-N
70642
CC1=CC2=C(N1)C=CC(=C2)OC
C10H11NO
MFCD00005620
1h-indole, 5-methoxy-2-methyl, 2-methyl-5-methoxy indole, 2-methyl-5-methoxyindole, 2-methyl-5-methoxylindole, 5-methoxy-2 methylindole, 5-methoxy-2-methylindole, acmc-1buiu, indole, 5-methoxy-2-methyl, maybridge1_002343, pubchem7236
5-methoxy-2-methyl-1H-indole
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 214-066-5

SDS Documents