missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Methoxy-2-methylbenzothiazole, TCI America™

$131.39 - $452.23

Chemical Identifiers

CAS 2941-69-7
Molecular Formula C9H9NOS
Molecular Weight (g/mol) 179.237
MDL Number MFCD00082664
InChI Key SAQMNBWVOKYKPZ-UHFFFAOYSA-N
Synonym 5-methoxy-2-methylbenzothiazole, 5-methoxy-2-methylbenzo d thiazole, benzothiazole, 5-methoxy-2-methyl, 2-methyl-5-methoxybenzothiazole, 2-methyl-5-methoxy-benzothiazole, acmc-209h8o, cambridge id 5150005, benzothiazole,5-methoxy-2-methyl, 2-methyl-5-methoxy-1,3-benzothiazole, 2-methyl-5-methoxy-1, 3-benzothiazole
PubChem CID 76252
IUPAC Name 5-methoxy-2-methyl-1,3-benzothiazole
SMILES CC1=NC2=C(S1)C=CC(=C2)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M12815G View Documents TCI America
M12815G
5g Each for $131.39
 
M128125G View Documents TCI America
M128125G
25g Each for $452.23
 
Specifications

Chemical Identifiers

2941-69-7
179.237
SAQMNBWVOKYKPZ-UHFFFAOYSA-N
76252
CC1=NC2=C(S1)C=CC(=C2)OC
C9H9NOS
MFCD00082664
5-methoxy-2-methylbenzothiazole, 5-methoxy-2-methylbenzo d thiazole, benzothiazole, 5-methoxy-2-methyl, 2-methyl-5-methoxybenzothiazole, 2-methyl-5-methoxy-benzothiazole, acmc-209h8o, cambridge id 5150005, benzothiazole,5-methoxy-2-methyl, 2-methyl-5-methoxy-1,3-benzothiazole, 2-methyl-5-methoxy-1, 3-benzothiazole
5-methoxy-2-methyl-1,3-benzothiazole

Specifications

135°C
37°C
2941-69-7
MFCD00082664
SAQMNBWVOKYKPZ-UHFFFAOYSA-N
5-methoxy-2-methyl-1,3-benzothiazole
76252
Lump at 20°C
5-Methoxy-2-methylbenzothiazole
White-Yellow
5g
C9H9NOS
5-methoxy-2-methylbenzothiazole, 5-methoxy-2-methylbenzo d thiazole, benzothiazole, 5-methoxy-2-methyl, 2-methyl-5-methoxybenzothiazole, 2-methyl-5-methoxy-benzothiazole, acmc-209h8o, cambridge id 5150005, benzothiazole,5-methoxy-2-methyl, 2-methyl-5-methoxy-1,3-benzothiazole, 2-methyl-5-methoxy-1, 3-benzothiazole
CC1=NC2=C(S1)C=CC(=C2)OC
179.237
179.24
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : Yes