Please login to your online account to display your discounted pricing

5-Methoxy-2-methylbenzothiazole 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC413840010

 View more versions of this product

Catalog No. AC413840010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Chunks
Formula Weight 179.24
Quantity 1g
Color Light brown-green semi-transparent
EINECS Number 220-929-7
TSCA TSCA
Melting Point 34° to 38°C
CAS Max % 100.0
Chemical Name or Material 5-Methoxy-2-methylbenzothiazole
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 98%
Assay Percent Range 98%
Molecular Formula C9H9NOS
MDL Number MFCD00082664
CAS 2941-69-7
CAS Min % 97.5

Specifications

Chemical Name or Material 5-Methoxy-2-methylbenzothiazole
Name Note 98%
CAS 2941-69-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H9NOS
Formula Weight 179.24
MDL Number MFCD00082664