Please login to your online account to display your discounted pricing

5-Methoxy-2-methylbenzothiazole 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 5-Methoxy-2-methylbenzothiazole
Name Note 98%
CAS 2941-69-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H9NOS
Formula Weight 179.24
MDL Number MFCD00082664
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC413840010

  SDS
thermo fisher scientific
AC413840010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Chunks
Formula Weight 179.24
Quantity 1g
Color Light brown-green semi-transparent
EINECS Number 220-929-7
TSCA TSCA
Melting Point 34° to 38°C
CAS Max % 100.0
CAS Min % 97.5
Packaging Glass bottle
Infrared Spectrum Authentic
Molecular Formula C9H9NOS
MDL Number MFCD00082664
Chemical Name or Material 5-Methoxy-2-methylbenzothiazole
Name Note 98%
Assay Percent Range 98%
CAS 2941-69-7