missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Methoxy-2-methylbenzoselenazole 96.0+%, TCI America™

$46.08 - $133.31

Chemical Identifiers

CAS 2946-17-0
Molecular Formula C9H9NOSe
Molecular Weight (g/mol) 226.148
MDL Number MFCD00005558
InChI Key LVIYTMFDCXRSDL-UHFFFAOYSA-N
Synonym #, 2-methyl-5-methoxybenzoselenazole, 5-methoxy-2-methylbenzo d 1,3 selenazole, 5-methoxy-2-methylbenzo d 1,3-selenazole, 5-methoxy-2-methylbenzo d 1,3-selenazoline, 5-methoxy-2-methylbenzoselenazole, 5-methoxy-2-methylbenzselenazole, benzoselenazole, 5-methoxy-2-methyl
PubChem CID 76274
IUPAC Name 5-methoxy-2-methyl-1,3-benzoselenazole
SMILES CC1=NC2=C([Se]1)C=CC(=C2)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M19001G View Documents Tci America
M19001G
1g Each for $46.08
 
M19005G View Documents Tci America
M19005G
5g Each for $133.31
 
Specifications

Chemical Identifiers

2946-17-0
226.148
LVIYTMFDCXRSDL-UHFFFAOYSA-N
76274
CC1=NC2=C([Se]1)C=CC(=C2)OC
C9H9NOSe
MFCD00005558
#, 2-methyl-5-methoxybenzoselenazole, 5-methoxy-2-methylbenzo d 1,3 selenazole, 5-methoxy-2-methylbenzo d 1,3-selenazole, 5-methoxy-2-methylbenzo d 1,3-selenazoline, 5-methoxy-2-methylbenzoselenazole, 5-methoxy-2-methylbenzselenazole, benzoselenazole, 5-methoxy-2-methyl
5-methoxy-2-methyl-1,3-benzoselenazole

Specifications

142°C
1g
C9H9NOSe
3283
LVIYTMFDCXRSDL-UHFFFAOYSA-N
5-methoxy-2-methyl-1,3-benzoselenazole
76274
Crystal-Lump at 20°C
5-Methoxy-2-methylbenzoselenazole
Yellow
2946-17-0
MFCD00005558
#, 2-methyl-5-methoxybenzoselenazole, 5-methoxy-2-methylbenzo d 1,3 selenazole, 5-methoxy-2-methylbenzo d 1,3-selenazole, 5-methoxy-2-methylbenzo d 1,3-selenazoline, 5-methoxy-2-methylbenzoselenazole, 5-methoxy-2-methylbenzselenazole, benzoselenazole, 5-methoxy-2-methyl
CC1=NC2=C([Se]1)C=CC(=C2)OC
226.148
226.14
≥96.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0498

TSCA : Yes