Please login to your online account to display your discounted pricing

5-Methoxy-2-methylaniline, 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC398240010

 View more versions of this product

Catalog No. AC398240010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Color Off-white to pale grey to yellow
Assay ≥96.0% (GC)
Formula Weight 137.18
Physical Form Powder
Quantity 1g
Melting Point 42° to 46°C
CAS Min % 96.0
CAS Max % 100.0
Linear Formula CH3OC6H3(CH3)NH2
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
Harmful in contact with skin.
Assay Percent Range 97%
Chemical Name or Material 5-Methoxy-2-methylaniline
MDL Number MFCD00075057
CAS 50868-72-9
Molecular Formula C8H11NO
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 5-Methoxy-2-methylaniline
CAS 50868-72-9
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0% (GC)
Assay Percent Range 97%
Molecular Formula C8H11NO
Linear Formula CH3OC6H3(CH3)NH2
Formula Weight 137.18
MDL Number MFCD00075057
Accessories

Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
Harmful in contact with skin.

SDS & Safety Documents