missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Methoxy-2-methylaniline, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 398240050
Specifications
5-Methoxy-2-methylaniline | |
96.0 | |
113°C | |
97% | |
C8H11NO | |
MFCD00075057 | |
5-methoxy-o-toluidine, 2-amino-4-methoxytoluene, benzenamine, 5-methoxy-2-methyl, 5-methoxy-2-methylbenzenamine, 2-methyl-5-ethoxyaniline, 5-methoxy-2-methylphenylamine, 6-methyl-m-anisidine, 3-amino-4-methylanisole 5-methoxy-2-methylaniline, pubchem19827, acmc-1awic | |
COC1=CC=C(C)C(N)=C1 | |
137.18 | |
137.18 |
50868-72-9 | |
100.0 | |
Authentic | |
Glass bottle | |
CH3OC6H3(CH3)NH2 | |
5g | |
RPJXLEZOFUNGNZ-UHFFFAOYSA-N | |
5-methoxy-2-methylaniline | |
99500 | |
97% |
Chemical Identifiers
50868-72-9 | |
137.18 | |
RPJXLEZOFUNGNZ-UHFFFAOYSA-N | |
99500 | |
COC1=CC=C(C)C(N)=C1 |
C8H11NO | |
MFCD00075057 | |
5-methoxy-o-toluidine, 2-amino-4-methoxytoluene, benzenamine, 5-methoxy-2-methyl, 5-methoxy-2-methylbenzenamine, 2-methyl-5-ethoxyaniline, 5-methoxy-2-methylphenylamine, 6-methyl-m-anisidine, 3-amino-4-methylanisole 5-methoxy-2-methylaniline, pubchem19827, acmc-1awic | |
5-methoxy-2-methylaniline |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
EINECSNumber : 256-816-4
RUO â Research Use Only