missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Methoxy-2-methyl-3-indoleacetic acid, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 147510050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
5-Methoxy-2-methyl-3-indoleacetic acid | |
2882-15-7 | |
100.0 | |
98% | |
C12H13NO3 | |
5g | |
Solubility in water: insoluble in water | |
CC1=C(C2=C(N1)C=CC(=C2)OC)CC(=O)O | |
219.24 | |
219.24 |
98% | |
97.5 | |
Authentic | |
Glass bottle | |
MFCD00005618 | |
5-methoxy-2-methyl-3-indoleacetic acid, 5 miaa, 2-5-methoxy-2-methyl-1h-indol-3-yl acetic acid, 5-methoxy-2-methyl-1h-indol-3-yl acetic acid, 1h-indole-3-acetic acid, 5-methoxy-2-methyl, n-deschlorobenzoyl indomethacin, 5-methoxy-2-methylindole-3-acetic acid, 5-methoxy-methylindoleacetic acid, deschlorobenzoyl indomethacin, 5-methoxy-2-methyl indole acetic acid | |
TXWGINUZLBAKDF-UHFFFAOYSA-N | |
2-(5-methoxy-2-methyl-1H-indol-3-yl)acetic acid | |
76151 | |
98% |
Chemical Identifiers
2882-15-7 | |
219.24 | |
TXWGINUZLBAKDF-UHFFFAOYSA-N | |
76151 | |
CC1=C(C2=C(N1)C=CC(=C2)OC)CC(=O)O |
C12H13NO3 | |
MFCD00005618 | |
5-methoxy-2-methyl-3-indoleacetic acid, 5 miaa, 2-5-methoxy-2-methyl-1h-indol-3-yl acetic acid, 5-methoxy-2-methyl-1h-indol-3-yl acetic acid, 1h-indole-3-acetic acid, 5-methoxy-2-methyl, n-deschlorobenzoyl indomethacin, 5-methoxy-2-methylindole-3-acetic acid, 5-methoxy-methylindoleacetic acid, deschlorobenzoyl indomethacin, 5-methoxy-2-methyl indole acetic acid | |
2-(5-methoxy-2-methyl-1H-indol-3-yl)acetic acid |
Safety and Handling
EINECSNumber : 220-734-7
RUO â Research Use Only