missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Methoxy-2-mercaptobenzimidazole, 99+%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 275370050
Specifications
(1% soln. in methanol) (1 cm cell) no other absorption observed from 360 to 600nm | |
Authentic | |
Glass bottle | |
Brown-Yellow to White | |
5g | |
37052-78-1 | |
100.0 | |
C8H8N2OS | |
1,3-dihydro-5-methoxy-2h-benzimidazole-2-thione, 2-mercapto-5-methoxy-1h-benzimidazole, 2-mercapto-5-methoxybenzimidazole, 5-methoxy-1h-1,3-benzodiazole-2-thiol, 5-methoxy-1h-benzimidazole-2-thiol, 5-methoxy-1h-benzo d imidazole-2-thiol, 5-methoxy-1h-benzoimidazole-2-thiol, 5-methoxy-2-benzimidazolethiol, 5-methoxy-2-mercaptobenzimidazole, 5-methoxybenzimidazole-2-thiol | |
KOFBRZWVWJCLGM-UHFFFAOYSA-N | |
5-methoxy-1,3-dihydrobenzimidazole-2-thione | |
665603 | |
Powder |
5-Methoxy-2-mercaptobenzimidazole | |
0.5% max. | |
1% max. | |
255°C to 262°C | |
99+% | |
99.0 | |
99% min. (HPLC) | |
MFCD00134581 | |
Solubility in water: insoluble. Other solubilities: soluble in sodium hydroxide solution | |
COC1=CC2=C(C=C1)NC(=S)N2 | |
180.23 | |
180.23 | |
99+% |
Chemical Identifiers
37052-78-1 | |
180.23 | |
KOFBRZWVWJCLGM-UHFFFAOYSA-N | |
665603 | |
COC1=CC2=C(C=C1)NC(=S)N2 |
C8H8N2OS | |
MFCD00134581 | |
1,3-dihydro-5-methoxy-2h-benzimidazole-2-thione, 2-mercapto-5-methoxy-1h-benzimidazole, 2-mercapto-5-methoxybenzimidazole, 5-methoxy-1h-1,3-benzodiazole-2-thiol, 5-methoxy-1h-benzimidazole-2-thiol, 5-methoxy-1h-benzo d imidazole-2-thiol, 5-methoxy-1h-benzoimidazole-2-thiol, 5-methoxy-2-benzimidazolethiol, 5-methoxy-2-mercaptobenzimidazole, 5-methoxybenzimidazole-2-thiol | |
5-methoxy-1,3-dihydrobenzimidazole-2-thione |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective cloth
GHS Signal Word: Warning
EINECSNumber : 253-326-2