Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Methoxy-1-tetralone, 97%

$40.65 - $140.39

Chemical Identifiers

Chemical Name or Material 5-Methoxy-1-tetralone
CAS 33892-75-0
Molecular Formula C11H12O2
Formula Weight 176.22
MDL Number MFCD00001692
Beilstein 2047383
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1512903

  SDS
alfa aesar™
L1512903
1g Each for $40.65

AAL1512906

  SDS
alfa aesar™
L1512906
5g Each for $140.39
Description & Specifications

Specifications

Formula Weight 176.22
Chemical Name or Material 5-Methoxy-1-tetralone
Assay 97%
Molecular Formula C11H12O2
MDL Number MFCD00001692
CAS 33892-75-0
Beilstein 2047383
Melting Point 86° to 91°C
RTECS Number QK4800000
Boiling Point 160° to 162°C (7mmHg)
Quantity 1g