Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Methoxy-1-methylindole-3-carboxaldehyde, 98%

$82.97 - $226.27

Chemical Identifiers

Chemical Name or Material 5-Methoxy-1-methylindole-3-carboxaldehyde
CAS 39974-94-2
Molecular Formula C11H11NO2
Formula Weight 189.22
MDL Number MFCD01578324
Beilstein 1528827
Synonym 3-Formyl-5-methoxy-1-methylindole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1926903

 
alfa aesar™
L1926903
1g Each for $226.27

AAL19269MD

 
alfa aesar™
L19269MD
250mg Each for $82.97
Description & Specifications

Specifications

Formula Weight 189.22
Chemical Name or Material 5-Methoxy-1-methylindole-3-carboxaldehyde
Molecular Formula C11H11NO2
MDL Number MFCD01578324
Assay 98%
CAS 39974-94-2
Beilstein 1528827
Synonym 3-Formyl-5-methoxy-1-methylindole
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 133° to 136°C
Quantity 1g

Applications
5-Methoxy-1-methylindole-3-carboxaldehyde is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
It is insoluble in water.

Notes
Air Sensitive. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.