Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Methoxy-1-methyl-4-nitroindole-3-carboxaldehyde, 90+%

$244.09 - $827.22

Chemical Identifiers

Chemical Name or Material 5-Methoxy-1-methyl-4-nitroindole-3-carboxaldehyde
CAS 191846-76-1
Molecular Formula C11H10N2O4
Formula Weight 234.21
MDL Number MFCD03093098
Beilstein 7766233
Synonym 3-Formyl-5-methoxy-1-methyl-4-nitroindole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1927003

  SDS
alfa aesar™
L1927003
1g Each for $244.09

AAL1927006

  SDS
alfa aesar™
L1927006
5g Each for $827.22
Description & Specifications

Specifications

Formula Weight 234.21
Molecular Formula C11H10N2O4
MDL Number MFCD03093098
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Chemical Name or Material 5-Methoxy-1-methyl-4-nitroindole-3-carboxaldehyde
Assay ≥90%
CAS 191846-76-1
Synonym 3-Formyl-5-methoxy-1-methyl-4-nitroindole
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point ≈190°C
Quantity 1g
Beilstein 7766233
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.