missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Mercapto-3-phenyl-1,3,4-thiadiazole-2(3H)thione potassium salt

As a complexing reagent for Bi, Te, Se and other metal ions

Manufacturer:  Alfa Aesar™ 4421909

 View more versions of this product

Catalog No. AA4421909



Specifications

Specifications

5-Mercapto-3-phenyl-1,3,4-thiadiazole-2 (3H)thione potassium salt
10g
C8H5KN2S3
1,3,4-thiadiazolidine-2,5-dithione, 3-phenyl-, potassium salt
QSKDCVGFAPHSHX-UHFFFAOYSA-M
potassium;4-phenyl-5-sulfanylidene-1,3,4-thiadiazole-2-thiolate
2775614
Powder
Hygroscopic
6336-51-2
MFCD00014092
It is soluble in water.
C1=CC=C(C=C1)N2C(=S)SC(=N2)[S-].[K+]
264.42
264.43

Chemical Identifiers

6336-51-2
264.42
QSKDCVGFAPHSHX-UHFFFAOYSA-M
2775614
C1=CC=C(C=C1)N2C(=S)SC(=N2)[S-].[K+]
C8H5KN2S3
MFCD00014092
1,3,4-thiadiazolidine-2,5-dithione, 3-phenyl-, potassium salt
potassium;4-phenyl-5-sulfanylidene-1,3,4-thiadiazole-2-thiolate
Safety and Handling

Safety and Handling

GHS H Statement
H301-H332-H315-H319-H335
Toxic if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H332-H315-H319-H335

EINECSNumber : 228-718-1

TSCA : No

Recommended Storage : Ambient temperatures

Documents