missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Mercapto-1H-1,2,3-triazole Sodium Salt, TCI America™

Manufacturer:  TCI America M084725G

 View more versions of this product

Catalog No. M084725G



Specifications

Specifications

5-Mercapto-1H-1,2,3-triazole Sodium Salt
25g
C2H2N3NaS
sodium 1,2,3-triazole-5-thiolate, 5-mercapto-1,2,3-triazole monosodium salt, sodium 1h-1,2,3-triazol-5-ylsulfanide, 1h-1,2,3-triazole-5-thiol sodium salt, 1h-5-mercapto-1,2,3-triazole,sodium salt, tas:1h-1,2,3-triazole-4-thiol sodium salt, sodium 1h-1,2,3-triazole-4-thiolate, sodium 1h-1,2,3-triazole-5-thiolate, sodium 2h-1,2,3-triazole-4-thiolate, 1h-1,2,3-triazole-4-thiol sodium salt
C1=NNN=C1[S-].[Na+]
123.109
123.11
≥97.0% (T)
White-Yellow
59032-27-8
MFCD00044799
HHGMADGROXARPN-UHFFFAOYSA-M
sodium;2H-triazole-4-thiolate
23685427
Crystal-Powder at 20°C

Chemical Identifiers

59032-27-8
123.109
HHGMADGROXARPN-UHFFFAOYSA-M
23685427
C1=NNN=C1[S-].[Na+]
C2H2N3NaS
MFCD00044799
sodium 1,2,3-triazole-5-thiolate, 5-mercapto-1,2,3-triazole monosodium salt, sodium 1h-1,2,3-triazol-5-ylsulfanide, 1h-1,2,3-triazole-5-thiol sodium salt, 1h-5-mercapto-1,2,3-triazole,sodium salt, tas:1h-1,2,3-triazole-4-thiol sodium salt, sodium 1h-1,2,3-triazole-4-thiolate, sodium 1h-1,2,3-triazole-5-thiolate, sodium 2h-1,2,3-triazole-4-thiolate, 1h-1,2,3-triazole-4-thiol sodium salt
sodium;2H-triazole-4-thiolate
Safety and Handling

Safety and Handling

EINECSNumber : (5)-5827

TSCA : No

Documents