Please login to your online account to display your discounted pricing

Alfa Aesar™ (5-Mercapto-1,3,4-thiadiazol-2-ylthio)acetic acid, 96%

$76.50 - $283.23

Chemical Identifiers

Chemical Name or Material (5-Mercapto-1,3,4-thiadiazol-2-ylthio)acetic acid
CAS 53723-88-9
Molecular Formula C4H4N2O2S3
Formula Weight 208.28
MDL Number MFCD00129970
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2188203

  SDS
alfa aesar™
B2188203
1g Each for $76.50

AAB2188206

  SDS
alfa aesar™
B2188206
5g Each for $283.23
Description & Specifications

Specifications

Formula Weight 208.28
Chemical Name or Material (5-Mercapto-1,3,4-thiadiazol-2-ylthio)acetic acid
Assay 96%
Molecular Formula C4H4N2O2S3
MDL Number MFCD00129970
CAS 53723-88-9
Health Hazard 1 Warning
Melting Point 165° to 169°C
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.