Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Isopropyl-2-methoxybenzaldehyde, 97%

$66.15 - $213.96

Chemical Identifiers

Chemical Name or Material 5-Isopropyl-2-methoxybenzaldehyde
CAS 85902-68-7
Molecular Formula C11H14O2
Formula Weight 178.23
MDL Number MFCD03095324
Beilstein 5923572
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1473714

  SDS
alfa aesar™
L1473714
25g Each for $213.96

AAL1473706

  SDS
alfa aesar™
L1473706
5g Each for $66.15
Description & Specifications

Specifications

Formula Weight 178.23
MDL Number MFCD03095324
CAS 85902-68-7
Chemical Name or Material 5-Isopropyl-2-methoxybenzaldehyde
Assay 97%
Molecular Formula C11H14O2
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Health Hazard 1 Warning
Quantity 25g
Beilstein 5923572
Boiling Point 96°C (1mmHg)
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.