Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Isopropyl-1,3-cyclohexanedione, 99%, may contain upto ca. 2% water

$56.05 - $203.42

Chemical Identifiers

Chemical Name or Material 5-Isopropyl-1,3-cyclohexanedione
CAS 18456-87-6
Molecular Formula C9H14O2
Formula Weight 154.21
MDL Number MFCD00075263
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2743503

  SDS
alfa aesar™
H2743503
1g Each for $56.05

AAH2743506

  SDS
alfa aesar™
H2743506
5g Each for $203.42
Description & Specifications

Specifications

Formula Weight 154.21
Chemical Name or Material 5-Isopropyl-1,3-cyclohexanedione
Assay 99%
Assay Percent Notes may contain up to ≈2% water
Molecular Formula C9H14O2
MDL Number MFCD00075263
CAS 18456-87-6
Melting Point 78° to 82°C
Quantity 1g