Learn More
5-Isocyanato-1-methyl-1H-indole, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 884507-16-8 |
---|---|
Molecular Formula | C10H8N2O |
Molecular Weight (g/mol) | 172.19 |
MDL Number | MFCD09702358 |
InChI Key | RDQRMCJCROUHOF-UHFFFAOYSA-N |
Synonym | 1-methyl-1h-indole-5-yl isocyanate, 1-methylindol-5-isocyanate, 1h-indole, 5-isocyanato-1-methyl, 1h-indole,5-isocyanato-1-methyl, 5-isocyanato-1-methyl-1h-indole |
PubChem CID | 24229489 |
IUPAC Name | 5-isocyanato-1-methyl-1H-indole |
SMILES | CN1C=CC2=CC(=CC=C12)N=C=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
CC41406CB
|
Fisher Scientific Company
CC41406CB |
250mg | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC41406DA
|
Fisher Scientific Company
CC41406DA |
1g | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
884507-16-8 | |
172.19 | |
RDQRMCJCROUHOF-UHFFFAOYSA-N | |
24229489 | |
CN1C=CC2=CC(=CC=C12)N=C=O |
C10H8N2O | |
MFCD09702358 | |
1-methyl-1h-indole-5-yl isocyanate, 1-methylindol-5-isocyanate, 1h-indole, 5-isocyanato-1-methyl, 1h-indole,5-isocyanato-1-methyl, 5-isocyanato-1-methyl-1h-indole | |
5-isocyanato-1-methyl-1H-indole |
Specifications
884507-16-8 | |
100.0 | |
C10H8N2O | |
Amber Glass Bottle | |
250mg | |
CN1C=CC2=CC(=CC=C12)N=C=O | |
172.19 | |
172.19 | |
5-Isocyanato-1-methyl-1H-indole |
96.5 | |
97% | |
MFCD09702358 | |
1-methyl-1h-indole-5-yl isocyanate, 1-methylindol-5-isocyanate, 1h-indole, 5-isocyanato-1-methyl, 1h-indole,5-isocyanato-1-methyl, 5-isocyanato-1-methyl-1h-indole | |
RDQRMCJCROUHOF-UHFFFAOYSA-N | |
5-isocyanato-1-methylindole | |
24229489 | |
97% |
Safety and Handling
GHS H Statement:
Harmful if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger