missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Iodovanillin, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1516230

 View more versions of this product

Catalog No. AAA1516230



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5-Iodovanillin
181°C to 184°C
C8H7IO3
250 g
Air and light sensitive
FBBCSYADXYILEH-UHFFFAOYSA-N
4-hydroxy-3-iodo-5-methoxybenzaldehyde
79499
98%
5438-36-8
Irritating
MFCD00006941
2049762
5-iodovanillin, 5 iodovanillin, benzaldehyde, 4-hydroxy-3-iodo-5-methoxy, 5-iodo vanillin, 3-methoxy-4-hydroxy-5-iodobenzaldehyde, 5-iodovanilline, 5-iodovanilin, 5-iodo vaniline, acmc-1ao68
COC1=C(C(=CC(=C1)C=O)I)O
278.045
278.05

Chemical Identifiers

5438-36-8
278.045
FBBCSYADXYILEH-UHFFFAOYSA-N
79499
COC1=C(C(=CC(=C1)C=O)I)O
C8H7IO3
MFCD00006941
5-iodovanillin, 5 iodovanillin, benzaldehyde, 4-hydroxy-3-iodo-5-methoxy, 5-iodo vanillin, 3-methoxy-4-hydroxy-5-iodobenzaldehyde, 5-iodovanilline, 5-iodovanilin, 5-iodo vaniline, acmc-1ao68
4-hydroxy-3-iodo-5-methoxybenzaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 226-617-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only