Please login to your online account to display your discounted pricing

5-Iodosalicylic Acid 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 5-Iodosalicylic acid
Name Note 97%
CAS 119-30-2
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H5IO3
Formula Weight 264.01
MDL Number MFCD00002458
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC122421000

  SDS
fisher scientific acros
122421000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Powder
Formula Weight 264.01
Quantity 100g
Color White to slightly beige
Beilstein 10, 112
EINECS Number 204-313-5
RTECS Number VO4200000
Melting Point 196° to 201°C
CAS Max % 100.0
CAS Min % 96.0
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Name Note 97%
Assay Percent Range 97%
Molecular Formula C7H5IO3
MDL Number MFCD00002458
Chemical Name or Material 5-Iodosalicylic acid
CAS 119-30-2