missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Iodoisatin, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1498806
Description
5-Iodoisatin undergoes condensation reaction with phenol yields 5-iodophenolisatin, malonic acid yields 6-iodo-2-quinolone-4-carboxylic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Iodoisatin undergoes condensation reaction with phenol yields 5-iodophenolisatin, malonic acid yields 6-iodo-2-quinolone-4-carboxylic acid.
Solubility
Soluble acetone, acetic acid, ethanol. Insoluble in water.
Notes
Light Sensitive. Keep container tightly closed. Store away from oxidizing agents.
Specifications
5-Iodoisatin | |
∼276°C (decomposition) | |
MFCD00016899 | |
131248 | |
5-iodoisatin, 5-iodoindoline-2,3-dione, 5-iodo isatin, 5-iodoindole-2,3-dione, 5-iodo-2,3-dioxoindoline, 1h-indole-2,3-dione, 5-iodo, 5-iodo-2,3-dihydro-1h-indole-2,3-dione, 5-iodo-1h-benzo d azolidine-2,3-dione, 5-iodo-isatine, 5-iodo-isatin | |
OEUGDMOJQQLVAZ-UHFFFAOYSA-N | |
5-iodo-1H-indole-2,3-dione | |
88695 | |
97% |
20780-76-1 | |
C8H4INO2 | |
5 g | |
Light sensitive | |
Soluble acetone,acetic acid,ethanol. Insoluble in water. | |
C1=CC2=C(C=C1I)C(=O)C(=O)N2 | |
273.029 | |
273.03 |
Chemical Identifiers
20780-76-1 | |
273.029 | |
OEUGDMOJQQLVAZ-UHFFFAOYSA-N | |
88695 | |
C1=CC2=C(C=C1I)C(=O)C(=O)N2 |
C8H4INO2 | |
MFCD00016899 | |
5-iodoisatin, 5-iodoindoline-2,3-dione, 5-iodo isatin, 5-iodoindole-2,3-dione, 5-iodo-2,3-dioxoindoline, 1h-indole-2,3-dione, 5-iodo, 5-iodo-2,3-dihydro-1h-indole-2,3-dione, 5-iodo-1h-benzo d azolidine-2,3-dione, 5-iodo-isatine, 5-iodo-isatin | |
5-iodo-1H-indole-2,3-dione |
Safety and Handling
EINECSNumber : 244-035-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only