missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Iodoisatin, 97%

Manufacturer:  Alfa Aesar™ A1498822

 View more versions of this product

Catalog No. AAA1498822



Description

Description

Applications
5-Iodoisatin undergoes condensation reaction with phenol yields 5-iodophenolisatin, malonic acid yields 6-iodo-2-quinolone-4-carboxylic acid.

Solubility
Soluble acetone, acetic acid, ethanol. Insoluble in water.

Notes
Light Sensitive. Keep container tightly closed. Store away from oxidizing agents.
Specifications

Specifications

5-Iodoisatin
20780-76-1
C8H4INO2
131248
OEUGDMOJQQLVAZ-UHFFFAOYSA-N
5-iodo-1H-indole-2,3-dione
88695
97%
~276°C (decomposition)
100g
MFCD00016899
5-iodoisatin, 5-iodoindoline-2,3-dione, 5-iodo isatin, 5-iodoindole-2,3-dione, 5-iodo-2,3-dioxoindoline, 1h-indole-2,3-dione, 5-iodo, 5-iodo-2,3-dihydro-1h-indole-2,3-dione, 5-iodo-1h-benzo d azolidine-2,3-dione, 5-iodo-isatine, 5-iodo-isatin
C1=CC2=C(C=C1I)C(=O)C(=O)N2
273.029
273.03

Chemical Identifiers

20780-76-1
273.029
OEUGDMOJQQLVAZ-UHFFFAOYSA-N
88695
C1=CC2=C(C=C1I)C(=O)C(=O)N2
C8H4INO2
MFCD00016899
5-iodoisatin, 5-iodoindoline-2,3-dione, 5-iodo isatin, 5-iodoindole-2,3-dione, 5-iodo-2,3-dioxoindoline, 1h-indole-2,3-dione, 5-iodo, 5-iodo-2,3-dihydro-1h-indole-2,3-dione, 5-iodo-1h-benzo d azolidine-2,3-dione, 5-iodo-isatine, 5-iodo-isatin
5-iodo-1H-indole-2,3-dione
Documents