missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Iodoindole, 95%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC441590250
Specifications
5-Iodoindole | |
Brown to White | |
25g | |
94.0 | |
94% min. (HPLC) | |
1h-indole, 5-iodo, 5-iodanyl-1h-indole, 5-iodo indole, 5-iodoindole, acmc-1brxb, maybridge1_006499, zlchem 1099 | |
C1=CC2=C(C=CN2)C=C1I | |
243.04 | |
243.04 | |
95% |
Authentic | |
95°C to 99°C | |
16066-91-4 | |
100.0 | |
C8H6IN | |
TVQLYTUWUQMGMP-UHFFFAOYSA-N | |
5-iodo-1H-indole | |
2782313 | |
Solid or Crystalline Solid |
Chemical Identifiers
16066-91-4 | |
243.04 | |
1h-indole, 5-iodo, 5-iodanyl-1h-indole, 5-iodo indole, 5-iodoindole, acmc-1brxb, maybridge1_006499, zlchem 1099 | |
5-iodo-1H-indole |
C8H6IN | |
TVQLYTUWUQMGMP-UHFFFAOYSA-N | |
2782313 | |
C1=CC2=C(C=CN2)C=C1I |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
Warning