missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Iodo-o-toluic Acid, TCI America™

$32.08 - $111.53

Chemical Identifiers

CAS 54811-38-0
Molecular Formula C8H7IO2
Molecular Weight (g/mol) 262.046
MDL Number MFCD01570258
InChI Key WUBHOZQZSHGUFI-UHFFFAOYSA-N
Synonym 5-Iodo-2-methylbenzoic Acid
PubChem CID 621745
IUPAC Name 5-iodo-2-methylbenzoic acid
SMILES CC1=C(C=C(C=C1)I)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I09165G View Documents TCI America
I09165G
5g Each for $32.08
 
I091625G View Documents TCI America
I091625G
25g Each for $111.53
 
Specifications

Chemical Identifiers

54811-38-0
262.046
WUBHOZQZSHGUFI-UHFFFAOYSA-N
621745
CC1=C(C=C(C=C1)I)C(=O)O
C8H7IO2
MFCD01570258
5-Iodo-2-methylbenzoic Acid
5-iodo-2-methylbenzoic acid

Specifications

White
5g
C8H7IO2
5-Iodo-2-methylbenzoic Acid
CC1=C(C=C(C=C1)I)C(=O)O
262.046
262.05
≥98.0% (GC,T)
181°C
54811-38-0
MFCD01570258
WUBHOZQZSHGUFI-UHFFFAOYSA-N
5-iodo-2-methylbenzoic acid
621745
Crystal-Powder at 20°C
5-Iodo-o-toluic Acid
Safety and Handling

Safety and Handling

TSCA : No