missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Iodo-3-methyl-2-methylaminopyridine, 95%

$108.66 - $341.72

Chemical Identifiers

CAS 942206-33-9
Molecular Formula C7H9IN2
Formula Weight 248.07
MDL Number MFCD09037456
Synonym 5-Iodo-N,3-dimethyl-2-pyridinamine
Chemical Name or Material 5-Iodo-3-methyl-2-methylaminopyridine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2771803 SDS Alfa Aesar™
H2771803
1g Each for $341.72
AAH27718MD SDS Alfa Aesar™
H27718MD
250mg Each for $108.66
Specifications Description & Specifications

Chemical Identifiers

942206-33-9
248.07
5-Iodo-N,3-dimethyl-2-pyridinamine
C7H9IN2
MFCD09037456
5-Iodo-3-methyl-2-methylaminopyridine

Specifications

942206-33-9
248.07
70° to 73°C
1g
95%
C7H9IN2
MFCD09037456
5-Iodo-N,3-dimethyl-2-pyridinamine
5-Iodo-3-methyl-2-methylaminopyridine

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning