Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Iodo-2-methylaniline, 98%

Chemical Identifiers

Chemical Name or Material 5-Iodo-2-methylaniline
CAS 83863-33-6
Molecular Formula C7H8IN
Formula Weight 233.05
MDL Number MFCD00051531
UN Number UN2811
Beilstein 2078769
Synonym 2-Amino-4-iodotoluene; 5-Iodo-o-toluidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0740403

 
alfa aesar™
L0740403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0740406

 
alfa aesar™
L0740406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 233.05
Assay 98%
Molecular Formula C7H8IN
MDL Number MFCD00051531
CAS 83863-33-6
Chemical Name or Material 5-Iodo-2-methylaniline
UN Number UN2811
Beilstein 2078769
Synonym 2-Amino-4-iodotoluene; 5-Iodo-o-toluidine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Melting Point 46° to 49°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.