missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Iodo-2-methylaniline, 97%, ACROS Organics™
Chemical Identifiers
CAS | 83863-33-6 |
---|---|
Molecular Formula | C7H8IN |
Molecular Weight (g/mol) | 233.05 |
InChI Key | IOEHXNCBPIBDBZ-UHFFFAOYSA-N |
Synonym | #, 2-amino-4-iodotoluene, 2-methyl-5-iodoaniline, 5-iodo-2-methyl-aniline, 5-iodo-2-methyl-phenylamine, 5-iodo-2-methylbenzenamine, 5-iodo-2-methylphenylamine, 5-iodo-o-toluidine, benzenamine, 5-iodo-2-methyl, benzenamine,5-iodo-2-methyl |
PubChem CID | 522802 |
IUPAC Name | 5-iodo-2-methylaniline |
SMILES | CC1=C(C=C(C=C1)I)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC442430010
|
Thermo Fisher Scientific
AC442430010 |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC442430050
|
Thermo Fisher Scientific
AC442430050 |
5g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
83863-33-6 | |
233.05 | |
#, 2-amino-4-iodotoluene, 2-methyl-5-iodoaniline, 5-iodo-2-methyl-aniline, 5-iodo-2-methyl-phenylamine, 5-iodo-2-methylbenzenamine, 5-iodo-2-methylphenylamine, 5-iodo-o-toluidine, benzenamine, 5-iodo-2-methyl, benzenamine,5-iodo-2-methyl | |
5-iodo-2-methylaniline |
C7H8IN | |
IOEHXNCBPIBDBZ-UHFFFAOYSA-N | |
522802 | |
CC1=C(C=C(C=C1)I)N |
Specifications
>110°C | |
48°C to 50°C | |
83863-33-6 | |
C7H8IN | |
IOEHXNCBPIBDBZ-UHFFFAOYSA-N | |
5-iodo-2-methylaniline | |
522802 | |
97% |
Glass Bottle | |
1g | |
96% min. (GC) | |
#, 2-amino-4-iodotoluene, 2-methyl-5-iodoaniline, 5-iodo-2-methyl-aniline, 5-iodo-2-methyl-phenylamine, 5-iodo-2-methylbenzenamine, 5-iodo-2-methylphenylamine, 5-iodo-o-toluidine, benzenamine, 5-iodo-2-methyl, benzenamine,5-iodo-2-methyl | |
CC1=C(C=C(C=C1)I)N | |
233.05 | |
233.05 | |
5-Iodo-2-methylaniline |
Safety and Handling
GHS Signal Word: Danger
EINECSNumber : 281-094-2