missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Iodo-2-methoxypyridine, TCI America™

$128.34 - $436.95

Chemical Identifiers

CAS 13472-61-2
Molecular Formula C6H6INO
Molecular Weight (g/mol) 235.024
MDL Number MFCD07781180
InChI Key NTXRNCUPGYOZCN-UHFFFAOYSA-N
Synonym 2-methoxy-5-iodopyridine, pyridine, 5-iodo-2-methoxy, pyridine,5-iodo-2-methoxy, pubchem6606, acmc-1c1li, 5-iodanyl-2-methoxy-pyridine
PubChem CID 23423786
IUPAC Name 5-iodo-2-methoxypyridine
SMILES COC1=NC=C(C=C1)I
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I09255G View Documents TCI America
I09255G
5g Each for $436.95
 
I09251G View Documents TCI America
I09251G
1g Each for $128.34
 
Specifications

Chemical Identifiers

13472-61-2
235.024
NTXRNCUPGYOZCN-UHFFFAOYSA-N
23423786
COC1=NC=C(C=C1)I
C6H6INO
MFCD07781180
2-methoxy-5-iodopyridine, pyridine, 5-iodo-2-methoxy, pyridine,5-iodo-2-methoxy, pubchem6606, acmc-1c1li, 5-iodanyl-2-methoxy-pyridine
5-iodo-2-methoxypyridine

Specifications

74°C
5g
C6H6INO
2-methoxy-5-iodopyridine, pyridine, 5-iodo-2-methoxy, pyridine,5-iodo-2-methoxy, pubchem6606, acmc-1c1li, 5-iodanyl-2-methoxy-pyridine
COC1=NC=C(C=C1)I
235.024
235.02
≥98.0% (GC)
Undesignated
13472-61-2
MFCD07781180
NTXRNCUPGYOZCN-UHFFFAOYSA-N
5-iodo-2-methoxypyridine
23423786
Clear Liquid at 20°C
5-Iodo-2-methoxypyridine
Safety and Handling

Safety and Handling

TSCA : No