Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Iodo-2-methoxypyridine, 98%

$186.84 - $827.22

Chemical Identifiers

Chemical Name or Material 5-Iodo-2-methoxypyridine
CAS 13472-61-2
Molecular Formula C6H6INO
Formula Weight 235.03
MDL Number MFCD07781180
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5013503

  SDS
alfa aesar™
H5013503
1g Each for $186.84

AAH5013506

  SDS
alfa aesar™
H5013506
5g Each for $827.22
Description & Specifications

Specifications

Formula Weight 235.03
Assay 98%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD07781180
Molecular Formula C6H6INO
CAS 13472-61-2
Chemical Name or Material 5-Iodo-2-methoxypyridine
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.