missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Iodo-2-methoxypyridine, 97%, ACROS Organics™

Chemical Identifiers

CAS 13472-61-2
Molecular Formula C6H6INO
Molecular Weight (g/mol) 235.03
InChI Key NTXRNCUPGYOZCN-UHFFFAOYSA-N
Synonym 2-methoxy-5-iodopyridine, 5-iodanyl-2-methoxy-pyridine, acmc-1c1li, pubchem6606, pyridine, 5-iodo-2-methoxy, pyridine,5-iodo-2-methoxy
PubChem CID 23423786
IUPAC Name 5-iodo-2-methoxypyridine
SMILES COC1=NC=C(C=C1)I
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC438930010
SDS View Documents
Thermo Fisher Scientific
AC438930010
1g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC438930050
SDS View Documents
Thermo Fisher Scientific
AC438930050
5g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

13472-61-2
235.03
2-methoxy-5-iodopyridine, 5-iodanyl-2-methoxy-pyridine, acmc-1c1li, pubchem6606, pyridine, 5-iodo-2-methoxy, pyridine,5-iodo-2-methoxy
5-iodo-2-methoxypyridine
C6H6INO
NTXRNCUPGYOZCN-UHFFFAOYSA-N
23423786
COC1=NC=C(C=C1)I

Specifications

Authentic
106°C to 108°C (30.0 Torr)
1g
96.0
96% min. (GC)
2-methoxy-5-iodopyridine, 5-iodanyl-2-methoxy-pyridine, acmc-1c1li, pubchem6606, pyridine, 5-iodo-2-methoxy, pyridine,5-iodo-2-methoxy
COC1=NC=C(C=C1)I
235.03
235.03
97%
Glass bottle
Colorless to Yellow
13472-61-2
100.0
C6H6INO
NTXRNCUPGYOZCN-UHFFFAOYSA-N
5-iodo-2-methoxypyridine
23423786
Liquid
5-Iodo-2-methoxypyridine
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning