missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Iodo-2-methoxypyridine, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 438930050
Specifications
5-Iodo-2-methoxypyridine | |
96.0 | |
106°C to 108°C (30.0 Torr) | |
96% min. (GC) | |
C6H6INO | |
2-methoxy-5-iodopyridine, pyridine, 5-iodo-2-methoxy, pyridine,5-iodo-2-methoxy, pubchem6606, acmc-1c1li, 5-iodanyl-2-methoxy-pyridine | |
COC1=NC=C(C=C1)I | |
235.03 | |
235.03 |
13472-61-2 | |
100.0 | |
Authentic | |
Glass bottle | |
5g | |
NTXRNCUPGYOZCN-UHFFFAOYSA-N | |
5-iodo-2-methoxypyridine | |
23423786 | |
97% |
Chemical Identifiers
13472-61-2 | |
235.03 | |
2-methoxy-5-iodopyridine, pyridine, 5-iodo-2-methoxy, pyridine,5-iodo-2-methoxy, pubchem6606, acmc-1c1li, 5-iodanyl-2-methoxy-pyridine | |
5-iodo-2-methoxypyridine |
C6H6INO | |
NTXRNCUPGYOZCN-UHFFFAOYSA-N | |
23423786 | |
COC1=NC=C(C=C1)I |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
RUO â Research Use Only