missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Iodo-2-methoxybenzaldehyde, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6632006
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
5-Iodo-2-methoxybenzaldehyde | |
140°C to 146°C | |
C8H7IO2 | |
5 g | |
2-methoxy-5-iodobenzaldehyde, 5-iodo-2-methoxy-benzaldehyde, pubchem1445, acmc-20am2z, benzaldehyde,5-iodo-2-methoxy, 5-iodo-2-methoxybenzaldehyde | |
COC1=CC=C(I)C=C1C=O | |
262.05 | |
262.04 | |
Solid |
42298-41-9 | |
White to Yellow | |
MFCD04128999 | |
Air and light sensitive | |
PIRKMHAREMCDPZ-UHFFFAOYSA-N | |
5-iodo-2-methoxybenzaldehyde | |
262240 | |
97% |
Chemical Identifiers
42298-41-9 | |
262.05 | |
PIRKMHAREMCDPZ-UHFFFAOYSA-N | |
262240 | |
COC1=CC=C(I)C=C1C=O |
C8H7IO2 | |
MFCD04128999 | |
2-methoxy-5-iodobenzaldehyde, 5-iodo-2-methoxy-benzaldehyde, pubchem1445, acmc-20am2z, benzaldehyde,5-iodo-2-methoxy, 5-iodo-2-methoxybenzaldehyde | |
5-iodo-2-methoxybenzaldehyde |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only