Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Iodo-2-methoxyaniline, 97%

$83.18 - $286.28

Chemical Identifiers

Chemical Name or Material 5-Iodo-2-methoxyaniline
CAS 77770-09-3
Molecular Formula C7H8INO
Formula Weight 249.05
MDL Number MFCD03095407
Synonym 2-Amino-4-iodoanisole; 5-Iodo-o-anisidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2555703

  SDS
alfa aesar™
B2555703
1g Each for $83.18

AAB2555706

  SDS
alfa aesar™
B2555706
5g Each for $286.28
Description & Specifications

Specifications

Formula Weight 249.05
Chemical Name or Material 5-Iodo-2-methoxyaniline
Assay 97%
Molecular Formula C7H8INO
MDL Number MFCD03095407
CAS 77770-09-3
Melting Point 86° to 87°C
Synonym 2-Amino-4-iodoanisole; 5-Iodo-o-anisidine
Quantity 1g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
5-Iodo-2-methoxyaniline is used as a pharmaceutical intermediate.

Solubility
Sparingly soluble in water 0.87 g/L @ 25°C.

Notes
Light sensitive. Store in dark. Store away from oxidizing agents, light.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.